Learn More
American Research Products Inc Rabbit intercellular adhesion
Catalog No. 5000330179
Encompass
Rabbit intercellular adhesion molecule 1 ICAM-1/CD54 ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-30179 Supplier American Research Products Inc Supplier No. CSBE06974RB
Rabbit intercellular adhesion molecule 1 ICAM-1/CD54 ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma Method Sandwich