Learn More
American Research Products Inc Rabbit Interleukin 1 IL 1 ELI
Catalog No. 5000329633
Encompass
Rabbit Interleukin 1 IL-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29633 Supplier American Research Products Inc Supplier No. CSBE06896RB
Rabbit Interleukin 1 IL-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich