Learn More
Biomatik Corporation Rabbit Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit
Supplier:  Biomatik Corporation EKU05159
Encompass_Preferred
Rabbit Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit for detecting IL1RA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26890. Gene ID: 100009558. Target Synonyms: IL1RN; IL1-RA; ICIL1RA; IL1ra3; IL1F3; IRAP;IL-1RA; Interleukin-1 Family Member 3; IL1 inhibitor; Anakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6393