Learn More
American Research Products Inc Rabbit Interleukin 10 IL 10 E
Catalog No. 5000328819
Encompass
Rabbit Interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28819 Supplier American Research Products Inc Supplier No. CSBE06897RB
Rabbit Interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich