Learn More
American Research Products Inc Rabbit interleukin 12 IL 12 P
Catalog No. 5000327274
Encompass
Rabbit interleukin 12 (IL-12/P70) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27274 Supplier American Research Products Inc Supplier No. CSBEQ027628RB
Rabbit interleukin 12 (IL-12/P70) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich