Learn More
American Research Products Inc Rabbit Interleukin 17 IL 17 E
Supplier:  American Research Products Inc CSBE08058RB
Encompass
Rabbit Interleukin 17 IL-17 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26261