Learn More
American Research Products Inc Rabbit Interleukin 18 IL 18 E
Catalog No. 5000328690
Encompass
Rabbit Interleukin 18 IL-18 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28690 Supplier American Research Products Inc Supplier No. CSBE13942RB
Rabbit Interleukin 18 IL-18 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma Method Sandwich