Learn More
American Research Products Inc Rabbit Interleukin 1b IL 1b E
Catalog No. 5000326286
Encompass
Rabbit Interleukin 1b IL-1b ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26286 Supplier American Research Products Inc Supplier No. CSBE06900RB
Rabbit Interleukin 1b IL-1b ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich