Learn More
American Research Products Inc Rabbit Interleukin 2 IL 2 ELI
Catalog No. 5000328710
Encompass
Rabbit Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28710 Supplier American Research Products Inc Supplier No. CSBE08745RB
Rabbit Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma Method Sandwich