Learn More
American Research Products Inc Rabbit Interleukin 4 IL 4 ELI
Catalog No. 5000326683
Encompass
Rabbit Interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 0 625 pg/mL-40 pg/mL Sensitivity 0 156 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26683 Supplier American Research Products Inc Supplier No. CSBE06902RB
Rabbit Interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 0 625 pg/mL-40 pg/mL Sensitivity 0 156 pg/mL Sample serum plasma tissue homogenates Method Sandwich