Learn More
American Research Products Inc Rabbit Interleukin 8 IL 8 ELI
Catalog No. 5000326684
Encompass
Rabbit Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26684 Supplier American Research Products Inc Supplier No. CSBE06905RB
Rabbit Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich