Learn More
American Research Products Inc Rabbit Krebs yon den Lundgen 6
Catalog No. 5000328694
Encompass
Rabbit Krebs yon den Lundgen-6 (KL-6) ELISA Kit from CUSABIO Detection Range 50 IU/mL-800 IU/mL Sensitivity 25 IU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28694 Supplier American Research Products Inc Supplier No. CSBE14112RB
Rabbit Krebs yon den Lundgen-6 (KL-6) ELISA Kit from CUSABIO Detection Range 50 IU/mL-800 IU/mL Sensitivity 25 IU/mL Sample serum plasma tissue homogenates Method Competitive