Learn More
American Research Products Inc Rabbit luteinizing hormone LH
Catalog No. 5000328368
Encompass
Rabbit luteinizing hormone (LH) ELISA kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28368 Supplier American Research Products Inc Supplier No. CSBE17780RB
Rabbit luteinizing hormone (LH) ELISA kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich