Learn More
Biomatik Corporation Rabbit Macrophage Inflammatory Protein 1 Beta (MIP1b) ELISA Kit
Supplier:  Biomatik Corporation EKU05751
Encompass_Preferred
Rabbit Macrophage Inflammatory Protein 1 Beta (MIP1b) ELISA Kit for detecting MIP1b in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46632. Gene ID: 100008984. Target Synonyms: CCL4; ACT2; G-26; LAG1; MIP1-B; SCYA4; Chemokine C-C-Motif Ligand 4; Small Inducible Cytokine A4; Homologous To Mouse Mip-1b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7395