Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 2 (MMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU05854
Encompass_Preferred
Rabbit Matrix Metalloproteinase 2 (MMP2) ELISA Kit for detecting MMP2 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50757. Gene ID: 100009000. Target Synonyms: CLG4; CLG4A; MMP-II; MONA; TBE-1; PEX; Gelatinase A; 72kDa Gelatinase; 72kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5083