Learn More
American Research Products Inc Rabbit MatriX Metalloproteinas
Catalog No. 5000328686
Encompass
Rabbit MatriX Metalloproteinase 13 MMP-13 ELISA Kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 44 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28686 Supplier American Research Products Inc Supplier No. CSBE14220RB
Rabbit MatriX Metalloproteinase 13 MMP-13 ELISA Kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 44 pg/mL Sample serum plasma tissue homogenates Method Sandwich