Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 2 (MMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09581
Encompass_Preferred
Rabbit Matrix Metalloproteinase 2 (MMP2) CLIA Kit for detecting MMP2 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P50757. Gene ID: 100009000. Target Synonyms: CLG4; CLG4A; MMP-II; MONA; TBE-1; PEX; Gelatinase A; 72kDa Gelatinase; 72kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8492