Learn More
American Research Products Inc Rabbit matriX metalloproteinas
Supplier:  American Research Products Inc CSBE06933RB
Encompass
Rabbit matriX metalloproteinase 9 MMP-9 ELISA Kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26288