Learn More
American Research Products Inc Rabbit matriX metalloproteinas
Supplier:  American Research Products Inc CSBE09744RB
Encompass
Rabbit matriX metalloproteinase 1 MMP-1 ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29340