Learn More
American Research Products Inc Rabbit matriX metalloproteinas
Catalog No. 5000326143
Encompass
Rabbit matriX metalloproteinase 2/Gelatinase A MMP-2 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26143 Supplier American Research Products Inc Supplier No. CSBE10373RB
Rabbit matriX metalloproteinase 2/Gelatinase A MMP-2 ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich