Learn More
Roche Diagnostics Rabbit Monoclonal Negative Control Ig
Supplier:  Roche Diagnostics 06683380001
Encompass
Rabbit Monoclonal Negative Control Ig is intended for laboratory use as a control for nonspecific binding of rabbit immunoglobulin in sections of formalin-fixed, paraffin-embedded tissue when stained on VENTANA BenchMark IHC/ISH platforms (BenchMark GX, BenchMark XT, and BenchMark ULTRA). This product should be interpreted by a qualified pathologist in conjunction with histological examination, relevant clinical information, and proper controls. This product is intended for in vitro diagnostic (IVD) use.
Catalog No. NC1705993