Learn More
American Research Products Inc Rabbit MyeloperoXidase MPO E
Catalog No. 5000329365
Encompass
Rabbit MyeloperoXidase (MPO) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29365 Supplier American Research Products Inc Supplier No. CSBE13689RB
Rabbit MyeloperoXidase (MPO) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive