Learn More
Biomatik Corporation Rabbit Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit
Supplier:  Biomatik Corporation EKU06311
Encompass_Preferred
Rabbit Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit for detecting NOS2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1SGE1. Target Synonyms: NOS2A; INOS; HEP-NOS; I-NOS; Hepatocytes Oxide Synthase; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5980