Learn More
Biomatik Corporation Rabbit N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit
Supplier:  Biomatik Corporation EKU11318
Encompass_Preferred
Rabbit N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 61.7-5,000pg/ml. Sensitivity: 27.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: G1TPG4. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5059