Learn More
Cayman Chemical RabbIt ntI-HA IgGDyLIght 1ea
SDP
Catalog No. 502951152
Encompass_Preferred
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to DyLight 488 Antibody:Rabbit polyclonal Anti-HA (YPYDVPDYA) IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Isotype: IgG Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays
Catalog No. 50-295-1152 Supplier Cayman Chemical Supplier No. 112501 EA
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to DyLight 488 Antibody:Rabbit polyclonal Anti-HA (YPYDVPDYA) IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Isotype: IgG Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays