Learn More
Cayman Chemical RabbIt ntI-HA IgGSureLIght 1ea
SDP
Catalog No. 502955511
Encompass_Preferred
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to SureLight APC Antibody: Rabbit polyclonal Anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening, Time-resolved fluorescence energy transfer
Catalog No. 50-295-5511 Supplier Cayman Chemical Supplier No. 134061 EA
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to SureLight APC Antibody: Rabbit polyclonal Anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening, Time-resolved fluorescence energy transfer