Learn More
Cayman Chemical RabbIt ntI-HA IgGSureLIght 1ea
Supplier: Cayman Chemical 134311 EA
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to SureLight™ R-PE, 100 μg Antibody: Rabbit polyclonal Anti-HA (YPYDVPDYA) IgG Dye: SureLight™ R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays