Learn More
Cayman Chemical RabbIt ntI-HA IgGSureLIght 1ea
SDP
Catalog No. 502951661
Encompass_Preferred
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to SureLight PerCP, 100 μg Antibody: Rabbit polyclonal Anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-1661 Supplier Cayman Chemical Supplier No. 112771 EA
Rabbit Anti-HA (Hemagglutinin) IgG conjugated to SureLight PerCP, 100 μg Antibody: Rabbit polyclonal Anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting