Learn More
Cayman Chemical RabbIt ntI-T7 IgGDyLIght 1ea
SDP
Catalog No. 502951135
Encompass_Preferred
Rabbit Anti-T7 IgG conjugated to DyLight 488 Antibody: Anti-T7-tag (MASMTGGQQMG) IgG Isotype: IgG Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy
Catalog No. 50-295-1135 Supplier Cayman Chemical Supplier No. 112511 EA
Rabbit Anti-T7 IgG conjugated to DyLight 488 Antibody: Anti-T7-tag (MASMTGGQQMG) IgG Isotype: IgG Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy