Learn More
Cayman Chemical RabbIt ntI-T7 IgGSureLIght 1ea
SDP
Catalog No. 502955546
Encompass_Preferred
Rabbit Anti-T7 IgG conjugated to SureLight™ R-PEAntibody: Anti-T7 tag (MASMTGGQQMG) IgGUses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-295-5546 Supplier Cayman Chemical Supplier No. 134331 EA
Rabbit Anti-T7 IgG conjugated to SureLight™ R-PEAntibody: Anti-T7 tag (MASMTGGQQMG) IgGUses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays