Learn More
Cayman Chemical RabbIt ntIHA IgG DyLIght 6 1ea
SDP
Catalog No. 502951154
Encompass_Preferred
Rabbit anti-HA (Hemagglutinin) IgG conjugated to DyLight 650 Antibody: Rabbit polyclonal anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Immunogen: YPYDVPDYA Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays
Catalog No. 50-295-1154 Supplier Cayman Chemical Supplier No. 112671 EA
Rabbit anti-HA (Hemagglutinin) IgG conjugated to DyLight 650 Antibody: Rabbit polyclonal anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Immunogen: YPYDVPDYA Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays