Learn More
Cayman Chemical RabbIt ntIT7 IgG DyLIght 5 1ea
SDP
Catalog No. 502951280
Encompass_Preferred
Rabbit anti-T7 IgG conjugated to DyLight 550 Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) Dye: DyLight 550 Isotype: IgG Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy
Catalog No. 50-295-1280 Supplier Cayman Chemical Supplier No. 112601 EA
Rabbit anti-T7 IgG conjugated to DyLight 550 Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) Dye: DyLight 550 Isotype: IgG Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and microscopy