Learn More
Cayman Chemical RabbIt ntIT7 IgG SureLIght 1ea
SDP
Catalog No. 502954736
Encompass_Preferred
Rabbit anti-T7 IgG conjugated to SureLight APC Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) IgG Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening
Catalog No. 50-295-4736 Supplier Cayman Chemical Supplier No. 134091 EA
Rabbit anti-T7 IgG conjugated to SureLight APC Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) IgG Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening