Learn More
Cayman Chemical RabbIt ntIT7 IgG SureLIght 1ea
SDP
Catalog No. 502951495
Encompass_Preferred
Rabbit anti-T7 IgG conjugated to SureLight PerCP Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-1495 Supplier Cayman Chemical Supplier No. 112791 EA
Rabbit anti-T7 IgG conjugated to SureLight PerCP Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting