Learn More
Cayman Chemical RabbIt ntIV5 IgG DyLIght 6 1ea
SDP
Catalog No. 502951293
Encompass_Preferred
Rabbit anti-V5 IgG conjugated to DyLight 650 Antibody: Rabbit anti-V5-tag (GKPIPNPLLGLDST) IgG Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, Microscopy
Catalog No. 50-295-1293 Supplier Cayman Chemical Supplier No. 112691 EA
Rabbit anti-V5 IgG conjugated to DyLight 650 Antibody: Rabbit anti-V5-tag (GKPIPNPLLGLDST) IgG Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, Microscopy