Learn More
Cayman Chemical RabbIt ntIV5 IgG SureLIght 1ea
SDP
Catalog No. 502954760
Encompass_Preferred
Rabbit anti-V5 IgG conjugated to SureLight R-PE Antibody: Rabbit anti-V5 tag (GKPIPNPLLGLDST) Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-295-4760 Supplier Cayman Chemical Supplier No. 134301 EA
Rabbit anti-V5 IgG conjugated to SureLight R-PE Antibody: Rabbit anti-V5 tag (GKPIPNPLLGLDST) Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays