Learn More
Biocare Medical, LLC Rabbit-on-Farma HRP-Polymer, 25ml
SDP
Catalog No. NC0964426
Encompass_Preferred
Biocares PromARKTM series introduces Rabbit-on-Farma HRP-Polymer detection. The Rabbit-on-Farma HRP-Polymer is formulated to detect rabbit polyclonal or monoclonal antibodies and is affinity purified for minimal cross-reactivity to cow, horse, pig and sheep IgG proteins. The advantages of this polymer technology are increased sensitivity, reduction of IHC steps (no avidin/biotin block or link/enzyme conjugates), minimal cross-reactivity to endogenous IgG that allows multiple species flexibility, and in most cases, tissues do not require a protein block.
Catalog No. NC0964426 Supplier Biocare Medical, LLC Supplier No. BRR4009H
Biocares PromARKTM series introduces Rabbit-on-Farma HRP-Polymer detection. The Rabbit-on-Farma HRP-Polymer is formulated to detect rabbit polyclonal or monoclonal antibodies and is affinity purified for minimal cross-reactivity to cow, horse, pig and sheep IgG proteins. The advantages of this polymer technology are increased sensitivity, reduction of IHC steps (no avidin/biotin block or link/enzyme conjugates), minimal cross-reactivity to endogenous IgG that allows multiple species flexibility, and in most cases, tissues do not require a protein block.