Learn More
American Research Products Inc Rabbit Osteocalcin bone gla pr
Catalog No. 5000329280
Encompass
Rabbit Osteocalcin/bone gla protein OT/BGP ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29280 Supplier American Research Products Inc Supplier No. CSBE14060RB
Rabbit Osteocalcin/bone gla protein OT/BGP ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich