Learn More
American Research Products Inc Rabbit oXidized low density li
Catalog No. 5000328811
Encompass
Rabbit oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 078 mol/mL-5 mol/mL Sensitivity 0 02 mol/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28811 Supplier American Research Products Inc Supplier No. CSBE06991RB
Rabbit oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 078 mol/mL-5 mol/mL Sensitivity 0 02 mol/mL Sample serum plasma tissue homogenates Method Sandwich