Learn More
American Research Products Inc Rabbit Parathyroid hormone PT
Catalog No. 5000329782
Encompass
Rabbit Parathyroid hormone PTH ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29782 Supplier American Research Products Inc Supplier No. CSBE13244RB
Rabbit Parathyroid hormone PTH ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma Method Sandwich