Learn More
Biomatik Corporation Rabbit Parathyroid Hormone (PTH) ELISA Kit
Supplier:  Biomatik Corporation EKU11939
Encompass_Preferred
Rabbit Parathyroid Hormone (PTH) ELISA Kit for detecting PTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.52pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: G1SNZ6. Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5546