Learn More
Biomatik Corporation Rabbit Pigment Epithelium Derived Factor (PEDF) ELISA Kit
Supplier:  Biomatik Corporation EKU11706
Encompass_Preferred
Rabbit Pigment Epithelium Derived Factor (PEDF) ELISA Kit for detecting PEDF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1SCK5. Gene ID: 100356583. Target Synonyms: SERPINF1; EPC1; PIG35; SDF3; Serpin F1; Cell proliferation-inducing gene 35; Serpin Peptidase Inhibitor,Clade F Member 1 (Alpha-2 Antiplasmin) ; Stromal Cell Derived Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6274