Learn More
American Research Products Inc Rabbit plasminogen actiVator i
Catalog No. 5000329796
Encompass
Rabbit plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29796 Supplier American Research Products Inc Supplier No. CSBE06978RB
Rabbit plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive