Learn More
American Research Products Inc Rabbit Platelet DeriVed Growth
Catalog No. 5000329565
Encompass
Rabbit Platelet-DeriVed Growth Factor AB (PDGF-AB) ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29565 Supplier American Research Products Inc Supplier No. CSBE15910RB
Rabbit Platelet-DeriVed Growth Factor AB (PDGF-AB) ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich