Learn More
Enzo Life Sciences Tau (human) polyclonal antibody (TAUY9) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLTA31190100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 12-27 (E12DHAGTpYGLGDRKDQG27) of human tau isoform 6 (2N4R) phosphorylated at Tyr18. UniProt: P10636. Source: Purified from rabbit serum. Species Reactivity: Human. Applications: IHC (FS), WB. Recommended dilutions: Immunohistochemistry (formalin-fixed tissue section, use of high temperature antigen retrieval technique may be required). Western Blot (1:3000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In PBS, pH 7.2, containing 0.1% sodium azide. Handling: Store unopened vial at -20°C until required for use. Avoid repeated freeze-thaw cycles.
Catalog No. NC0487354