Learn More
American Research Products Inc Rabbit Procalcitonin PCT ELI
Catalog No. 5000329016
Encompass
Rabbit Procalcitonin (PCT) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29016 Supplier American Research Products Inc Supplier No. CSBE12875RB
Rabbit Procalcitonin (PCT) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich