Learn More
American Research Products Inc Rabbit procollagen I N termina
Catalog No. 5000326958
Encompass
Rabbit procollagen I N-terminal peptide (PINP) ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26958 Supplier American Research Products Inc Supplier No. CSBE17085RB
Rabbit procollagen I N-terminal peptide (PINP) ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich