Learn More
American Research Products Inc Rabbit prothrombin fragment 1
Catalog No. 5000327506
Encompass
Rabbit prothrombin fragment 1 2 (F1 2) ELISA kit from CUSABIO Detection Range 0 312 pmol/mL-20 pmol/mL Sensitivity 0 078 pmol/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27506 Supplier American Research Products Inc Supplier No. CSBEQ027672RB
Rabbit prothrombin fragment 1 2 (F1 2) ELISA kit from CUSABIO Detection Range 0 312 pmol/mL-20 pmol/mL Sensitivity 0 078 pmol/mL Sample serum plasma tissue homogenates Method Sandwich