Learn More
Cayman Chemical RabbIt PRoxISME ProlIfRato
SDP
Catalog No. 502945176
Encompass_Preferred
INDIGO’s assay kits provide all-inclusive cell-based assay systems. In addition to Rabbit PPARδ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist GW0742, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-294-5176 Supplier Cayman Chemical Supplier No. 410383X32 WELL
INDIGO’s assay kits provide all-inclusive cell-based assay systems. In addition to Rabbit PPARδ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist GW0742, Luciferase Detection Reagent, and a cell culture-ready assay plate.