Learn More
Cayman Chemical RabbIt PRoxIsome ProlIfRato 96
SDP
Catalog No. 502945082
Encompass_Preferred
INDIGO’s assay kits provide all-inclusive cell-based assay systems. In addition to Rabbit PPARδ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist GW0742, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-294-5082 Supplier Cayman Chemical Supplier No. 4103896 WELL
INDIGO’s assay kits provide all-inclusive cell-based assay systems. In addition to Rabbit PPARδ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist GW0742, Luciferase Detection Reagent, and a cell culture-ready assay plate.